Assay Details
| Gene Symbol: Brf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024773.1 | NP_001019944.1 | 2 - 3 | 281 | 110 | |
| GenBank mRNA | BC097435.1 | - | 2 - 3 | 281 | 7455454 | 110 |
More Information
Updated Mapping Information
Set Membership: |
|

Brf2
Genomic Map