Assay Details
Gene Symbol: Prosc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107320.1 | NP_001100790.1 | 1 - 2 | 345 | 100 | |
XM_008771329.1 | XP_008769551.1 | 1 - 2 | 151 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
