Assay Details
| Gene Symbol: Batf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230953.2 | XP_006231015.1 | 3 - 4 | 389 | 59 | |
| XM_008760203.1 | XP_008758425.1 | 2 - 3 | 974 | 59 | ||
| XM_219527.7 | XP_219527.5 | 2 - 3 | 318 | 59 | ||
More Information
Set Membership: |
|

Batf2
Genomic Map