Assay Details
| Gene Symbol: Fkbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134428.1 | NP_001127900.1 | 2 - 3 | 260 | 87 | |
| NM_001134429.1 | NP_001127901.1 | 2 - 3 | 296 | 87 | ||
| XM_006230756.1 | XP_006230818.1 | 2 - 3 | 348 | 87 | ||
| XM_006230757.1 | XP_006230819.1 | 2 - 3 | 312 | 87 | ||
| XM_006230758.2 | XP_006230820.1 | 2 - 3 | 750 | 87 | ||
| GenBank mRNA | BI285619.1 | - | 4 - 5 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp2
Genomic Map