Assay Details
| Gene Symbol: Mtg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749002.3 | XP_003749050.1 | 6 - 7 | 563 | 76 | |
| XM_006230584.2 | XP_006230646.1 | 6 - 7 | 564 | 76 | ||
| GenBank mRNA | FQ212842.1 | - | 6 - 7 | 529 | 76 | |
More Information
Set Membership: |
|

Mtg1
Genomic Map