Assay Details
| Gene Symbol: Sdhaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008371.1 | NP_001008372.1 | 1 - 2 | 90 | 84 | |
| XM_006231043.2 | XP_006231105.1 | 1 - 2 | 145 | 84 | ||
| GenBank mRNA | BC076396.1 | - | 1 - 2 | 43 | 84 | |
| BC086542.1 | - | 1 - 2 | 90 | 7122954 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sdhaf2
Genomic Map