Assay Details
| Gene Symbol: Nudt22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199090.2 | NP_954521.1 | 2 - 3 | 601 | 131 | |
| XM_006230760.2 | XP_006230822.1 | 1 - 2 | 503 | 131 | ||
| GenBank mRNA | BC060593.1 | - | 2 - 3 | 602 | 6887766 | 131 |
| FQ212766.1 | - | 2 - 3 | 548 | 131 | ||
| FQ215165.1 | - | 2 - 3 | 529 | 131 | ||
| FQ219356.1 | - | 2 - 3 | 552 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nudt22
Genomic Map