Assay Details
| Gene Symbol: Bad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230896.2 | XP_006230958.1 | 3 - 4 | 1192 | 79 | |
| XM_006230897.2 | XP_006230959.1 | 2 - 3 | 370 | 79 | ||
| GenBank mRNA | AF279911.1 | - | 3 - 4 | 487 | 79 | |
More Information
Set Membership: |
|

Bad
Genomic Map