Assay Details
| Gene Symbol: LOC100360606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728842.4 | XP_002728888.3 | 4 - 5 | 1567 | 77 | |
| XM_006230445.2 | XP_006230507.1 | 5 - 6 | 1732 | 77 | ||
| XM_008759959.1 | XP_008758181.1 | 4 - 5 | 1819 | 77 | ||
| XM_008759960.1 | XP_008758182.1 | 5 - 6 | 1464 | 77 | ||
More Information
Set Membership: |
|

LOC100360606
Genomic Map