Assay Details
| Gene Symbol: Kif11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169112.1 | NP_001162583.1 | 6 - 7 | 884 | 100 | |
| GenBank mRNA | AF035955.1 | - | 3 - 4 | 392 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif11
Genomic Map