Assay Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031801.1 | NP_113989.1 | 9 - 10 | 1359 | 95 | |
| XM_006230582.2 | XP_006230644.1 | 9 - 10 | 2227 | 95 | ||
| XM_006230583.2 | XP_006230645.1 | 9 - 10 | 2227 | 95 | ||
| XM_008760011.1 | XP_008758233.1 | 9 - 10 | 1278 | 95 | ||
| XM_008760012.1 | XP_008758234.1 | 9 - 10 | 1278 | 95 | ||
| XM_008760013.1 | XP_008758235.1 | 9 - 10 | 1278 | 95 | ||
| XM_008760014.1 | XP_008758236.1 | 9 - 10 | 2243 | 95 | ||
| XM_008760015.1 | XP_008758237.1 | 4 - 5 | 871 | 95 | ||
| XM_008760016.1 | XP_008758238.1 | 4 - 5 | 871 | 95 | ||
| GenBank mRNA | AF055884.1 | - | 9 - 10 | 1359 | 95 | |
| U59659.1 | - | 9 - 10 | 1105 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Deaf1
Genomic Map