Assay Details
| Gene Symbol: Cdk2ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109498.1 | NP_001102968.1 | 2 - 3 | 265 | 106 | |
| XM_006230902.2 | XP_006230964.1 | 2 - 3 | 655 | 106 | ||
| XM_006230903.2 | XP_006230965.1 | 2 - 3 | 203 | 106 | ||
| GenBank mRNA | FQ234216.1 | - | 2 - 3 | 267 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk2ap2
Genomic Map