Assay Details
| Gene Symbol: Tmem179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109572.1 | NP_001103042.1 | 1 - 2 | 125 | 128 | |
| XM_006231001.2 | XP_006231063.1 | 1 - 2 | 1018 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem179b
Genomic Map