Assay Details
| Gene Symbol: Zfand5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106356.1 | NP_001099826.1 | 2 - 3 | 756 | 76 | |
| XM_008760297.1 | XP_008758519.1 | 4 - 5 | 955 | 76 | ||
| GenBank mRNA | BC168882.1 | - | 4 - 5 | 423 | 9026652 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfand5
Genomic Map