Assay Details
| Gene Symbol: Ccnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171992.4 | NP_741989.3 | 1 - 2 | 325 | 86 | |
| XM_008760168.1 | XP_008758390.1 | 1 - 2 | 500 | 86 | ||
| GenBank mRNA | AB256048.1 | - | 1 - 2 | 378 | 86 | |
| AM258963.2 | - | 1 - 2 | 378 | 86 | ||
| CA510842.1 | - | 1 - 2 | 326 | 86 | ||
| D14014.1 | - | 1 - 2 | 325 | 86 | ||
| FQ231654.1 | - | 1 - 2 | 338 | 86 | ||
| X75207.1 | - | 1 - 2 | 352 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB256048.1, AM258963.2 transcripts.
Set Membership: |
|

Ccnd1
Genomic Map