Assay Details
| Gene Symbol: LOC100910717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749096.3 | XP_003749144.1 | 25 - 26 | 3732 | 66 | |
| Gene Symbol: Tnks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107607.2 | NP_001101077.2 | 25 - 26 | 3282 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
metabolic process
protein polyubiquitination
telomere maintenance
protein ADP-ribosylation
multicellular organism growth
regulation of multicellular organism growth
protein localization to chromosome, telomeric region
protein auto-ADP-ribosylation
positive regulation of canonical Wnt signaling pathway
NAD+ ADP-ribosyltransferase activity
enzyme binding
metabolic process
protein polyubiquitination
telomere maintenance
protein ADP-ribosylation
multicellular organism growth
regulation of multicellular organism growth
protein localization to chromosome, telomeric region
protein auto-ADP-ribosylation
positive regulation of canonical Wnt signaling pathway
NAD+ ADP-ribosyltransferase activity
enzyme binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P572344 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P112149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23637 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P572344 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P112149 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P572344 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P112149 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P572344 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P112149 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P112149 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P572344 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58957 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10715024 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715017 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714997 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715018 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715015 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715016 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715000 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715020 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715001 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715014 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715004 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715023 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715022 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714998 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715012 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715011 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715002 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715021 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715013 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715019 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715010 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10714995 | - | - |
| Rat Genome 230 2.0 Array | 1398172_at | - | - |
| Rat Genome 230 2.0 Array | 1376999_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395047_at | - | - |
| Rat Genome 230 2.0 Array | 1393423_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374595_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376999_at | - | - |
| Rat Genome 230 2.0 Array | 1398172_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393423_at | - | - |
| Rat Genome 230 2.0 Array | 1374595_at | - | - |
| Rat Genome 230 2.0 Array | 1395047_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376999_at | - | - |
| Affymetrix HT Rat Focus Array | 1395047_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393423_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376999_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374595_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374595_at | - | - |
| Affymetrix HT Rat Focus Array | 1395047_at | - | - |
| Affymetrix HT Rat Focus Array | 1393423_at | - | - |
| Rat Expression Set 230 | 1376999_PM_at | - | - |
| Rat Expression Set 230 | 1376999_at | - | - |
| Rat Expression Set 230 | 1374595_PM_at | - | - |
| Rat Expression Set 230 | 1374595_at | - | - |
| Rat Expression Set 230 | 1393423_at | - | - |
| Rat Expression Set 230 | 1395047_PM_at | - | - |
| Rat Expression Set 230 | 1395047_at | - | - |
| Rat Expression Set 230 | 1393423_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10714995 | - | - |
| Rat Genome U34 Set | rc_AA892570_at | - | - |
| Rat Genome U34 Set | rc_AA963756_at | - | - |
| Rat Genome U34 Set | rc_AI180334_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21375845 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358674 | - | - |

LOC100910717
Genomic Map