Assay Details
| Gene Symbol: LOC100911519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760031.1 | XP_008758253.1 | 1 - 2 | 457 | 119 | |
| Gene Symbol: Pidd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759984.1 | XP_008758206.1 | 1 - 2 | 457 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008759984.1, XM_008760031.1 transcripts.
Set Membership: |
|

LOC100911519
Genomic Map