Assay Details
| Gene Symbol: Rbm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007014.2 | NP_001007015.2 | 1 - 2 | 103 | 100 | |
| XM_008760164.1 | XP_008758386.1 | 1 - 2 | 265 | 100 | ||
| XM_008760166.1 | XP_008758388.1 | 1 - 2 | 265 | 100 | ||
| GenBank mRNA | AY303541.1 | - | 1 - 2 | 68 | 100 | |
| BC086416.1 | - | 1 - 2 | 103 | 7130401 | 100 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY303541.1,BC086416.1,NM_001007014.2,XM_008760164.1,XM_008760166.1 transcripts.
Set Membership: |
|

Rbm4b
Genomic Map