Assay Details
| Gene Symbol: Rplp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030021.1 | NP_001025192.1 | 1 - 2 | 44 | 90 | |
| GenBank mRNA | BC099697.1 | - | 1 - 2 | 44 | 7934014 | 90 |
| FQ217138.1 | - | 1 - 2 | 57 | 90 | ||
| FQ217658.1 | - | 1 - 2 | 58 | 90 | ||
| FQ221181.1 | - | 1 - 2 | 57 | 90 | ||
| FQ221478.1 | - | 1 - 2 | 60 | 90 | ||
| FQ221584.1 | - | 1 - 2 | 56 | 90 | ||
| FQ222461.1 | - | 1 - 2 | 56 | 90 | ||
| FQ222498.1 | - | 1 - 2 | 57 | 90 | ||
| FQ223422.1 | - | 1 - 2 | 59 | 90 | ||
| FQ228678.1 | - | 1 - 2 | 90 | |||
| Gene Symbol: LOC100911575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749013.2 | XP_003749061.1 | 1 - 2 | 279 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC099697.1, NM_001030021.1, XM_003749013.2 transcripts.
Set Membership: |
|

Rplp2
Genomic Map