Assay Details
| Gene Symbol: Tle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019141.2 | NP_062014.2 | 13 - 14 | 1445 | 67 | |
| XM_006231128.2 | XP_006231190.1 | 14 - 15 | 2131 | 67 | ||
| XM_006231129.2 | XP_006231191.1 | 14 - 15 | 2128 | 67 | ||
| XM_008760286.1 | XP_008758508.1 | 13 - 14 | 2053 | 67 | ||
| GenBank mRNA | CO389810.1 | - | 2 - 3 | 282 | 67 | |
| L14463.1 | - | 13 - 14 | 1438 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
Wnt signaling pathway
negative regulation of transcription, DNA-templated
negative regulation of nucleic acid-templated transcription
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
repressing transcription factor binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
Wnt signaling pathway
negative regulation of transcription, DNA-templated
negative regulation of nucleic acid-templated transcription
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
repressing transcription factor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P208004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71025 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P208004 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P208004 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P208004 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P208004 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27766 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2796 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12554 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63659 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4983 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79539 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46555 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7496 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97099 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10729090 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729084 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729081 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729076 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729082 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729092 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729075 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729089 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729087 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729086 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729078 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729080 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729083 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729091 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729085 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729079 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729077 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10729074 | - | - |
| Rat Genome 230 2.0 Array | 1387495_at | - | - |
| Rat Genome 230 2.0 Array | 1387495_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387495_at | - | - |
| Affymetrix HT Rat Focus Array | 1387495_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100059295.rn.1 | TC0100007340.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059296.rn.1 | TC0100007340.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059299.rn.1 | TC0100007340.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059292.rn.1 | TC0100007340.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059294.rn.1 | TC0100007340.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059298.rn.1 | TC0100007340.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059301.rn.1 | TC0100007340.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059297.rn.1 | TC0100007340.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100059300.rn.1 | TC0100007340.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387495_PM_at | - | - |
| Rat Expression Set 230 | 1387495_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729074 | - | - |
| Rat Genome U34 Set | L14463_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22262479 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373497 | - | - |
| RatRef-12 Expression BeadChip | ILMN_68937 | - | - |

Tle4
Genomic Map