Assay Details
| Gene Symbol: Tcerg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130077.1 | NP_001123549.1 | 11 - 12 | 1527 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P013569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69275 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013569 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013569 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P013569 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P257758 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P450055 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10726443 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726447 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726445 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726452 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726451 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726442 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726453 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726444 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726446 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726440 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726441 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10726439 | - | - |
| Rat Genome 230 2.0 Array | 1375618_at | - | - |
| Rat Genome 230 2.0 Array | 1375618_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391919_at | - | - |
| Rat Genome 230 2.0 Array | 1391919_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391919_at | - | - |
| Affymetrix HT Rat Focus Array | 1391919_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100053987.rn.1 | TC0100006867.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375618_PM_at | - | - |
| Rat Expression Set 230 | 1375618_at | - | - |
| Rat Expression Set 230 | 1391919_at | - | - |
| Rat Expression Set 230 | 1391919_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726439 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22074439 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360949 | - | - |

Tcerg1l
Genomic Map