Assay Details
| Gene Symbol: Cyp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012542.2 | NP_036674.1 | 5 - 6 | 830 | 113 | |
| GenBank mRNA | J02852.1 | - | 5 - 6 | 809 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2a3
Genomic Map