Assay Details
| Gene Symbol: Bcl2l12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108480.1 | NP_001101950.1 | 2 - 3 | 270 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108480.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bcl2l12
Genomic Map