Assay Details
| Gene Symbol: Bax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017059.2 | NP_058755.2 | 5 - 6 | 559 | 63 | |
| GenBank mRNA | AF235993.1 | - | 4 - 5 | 924 | 63 | |
| U59184.1 | - | 2 - 3 | 248 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
neuron migration
leukocyte homeostasis
T cell homeostatic proliferation
B cell homeostasis
B cell apoptotic process
kidney development
release of cytochrome c from mitochondria
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
blood vessel remodeling
myeloid cell homeostasis
B cell negative selection
B cell homeostatic proliferation
positive regulation of B cell apoptotic process
glycosphingolipid metabolic process
regulation of nitrogen utilization
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
obsolete transformed cell apoptotic process
cellular response to DNA damage stimulus
inner mitochondrial membrane organization
outer mitochondrial membrane organization
germ cell development
spermatogenesis
nervous system development
brain development
sex differentiation
mitochondrial fusion
cell proliferation
negative regulation of cell proliferation
male gonad development
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
apoptotic mitochondrial changes
fertilization
response to wounding
response to toxic substance
response to salt stress
post-embryonic development
response to ionizing radiation
establishment or maintenance of transmembrane electrochemical gradient
response to gamma radiation
positive regulation of calcium ion transport into cytosol
hypothalamus development
cerebral cortex development
negative regulation of protein binding
positive regulation of protein oligomerization
negative regulation of endoplasmic reticulum calcium ion concentration
release of matrix enzymes from mitochondria
negative regulation of peptidyl-serine phosphorylation
regulation of mammary gland epithelial cell proliferation
glial cell apoptotic process
cellular response to UV
limb morphogenesis
ectopic germ cell programmed cell death
response to cocaine
odontogenesis of dentin-containing tooth
response to drug
positive regulation of apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
positive regulation of neuron apoptotic process
mitochondrial fragmentation involved in apoptotic process
development of secondary sexual characteristics
cellular respiration
retinal cell programmed cell death
response to copper ion
positive regulation of developmental pigmentation
negative regulation of fibroblast proliferation
spermatid differentiation
post-embryonic camera-type eye morphogenesis
response to axon injury
homeostasis of number of cells
homeostasis of number of cells within a tissue
protein oligomerization
protein homooligomerization
positive regulation of release of sequestered calcium ion into cytosol
neuron apoptotic process
response to corticosterone
regulation of cell cycle
regulation of mitochondrial membrane potential
Sertoli cell proliferation
retina development in camera-type eye
positive regulation of apoptotic process involved in mammary gland involution
vagina development
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
thymocyte apoptotic process
mitochondrion morphogenesis
cellular response to organic substance
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
intrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum unfolded protein response
regulation of mitochondrial membrane permeability involved in apoptotic process
positive regulation of mitochondrial membrane permeability involved in apoptotic process
apoptotic process involved in patterning of blood vessels
apoptotic process involved in embryonic digit morphogenesis
regulation of mitochondrial membrane permeability involved in programmed necrotic cell death
positive regulation of apoptotic DNA fragmentation
retinal cell apoptotic process
B cell receptor apoptotic signaling pathway
negative regulation of apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of intrinsic apoptotic signaling pathway
protein binding
lipid binding
channel activity
heat shock protein binding
protein complex binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
chaperone binding
BH domain binding
BH3 domain binding
ovarian follicle development
neuron migration
leukocyte homeostasis
T cell homeostatic proliferation
B cell homeostasis
B cell apoptotic process
kidney development
release of cytochrome c from mitochondria
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
blood vessel remodeling
myeloid cell homeostasis
B cell negative selection
B cell homeostatic proliferation
positive regulation of B cell apoptotic process
glycosphingolipid metabolic process
regulation of nitrogen utilization
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
obsolete transformed cell apoptotic process
cellular response to DNA damage stimulus
inner mitochondrial membrane organization
outer mitochondrial membrane organization
germ cell development
spermatogenesis
nervous system development
brain development
sex differentiation
mitochondrial fusion
cell proliferation
negative regulation of cell proliferation
male gonad development
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
apoptotic mitochondrial changes
fertilization
response to wounding
response to toxic substance
response to salt stress
post-embryonic development
response to ionizing radiation
establishment or maintenance of transmembrane electrochemical gradient
response to gamma radiation
positive regulation of calcium ion transport into cytosol
hypothalamus development
cerebral cortex development
negative regulation of protein binding
positive regulation of protein oligomerization
negative regulation of endoplasmic reticulum calcium ion concentration
release of matrix enzymes from mitochondria
negative regulation of peptidyl-serine phosphorylation
regulation of mammary gland epithelial cell proliferation
glial cell apoptotic process
cellular response to UV
limb morphogenesis
ectopic germ cell programmed cell death
response to cocaine
odontogenesis of dentin-containing tooth
response to drug
positive regulation of apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
positive regulation of neuron apoptotic process
mitochondrial fragmentation involved in apoptotic process
development of secondary sexual characteristics
cellular respiration
retinal cell programmed cell death
response to copper ion
positive regulation of developmental pigmentation
negative regulation of fibroblast proliferation
spermatid differentiation
post-embryonic camera-type eye morphogenesis
response to axon injury
homeostasis of number of cells
homeostasis of number of cells within a tissue
protein oligomerization
protein homooligomerization
positive regulation of release of sequestered calcium ion into cytosol
neuron apoptotic process
response to corticosterone
regulation of cell cycle
regulation of mitochondrial membrane potential
Sertoli cell proliferation
retina development in camera-type eye
positive regulation of apoptotic process involved in mammary gland involution
vagina development
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
thymocyte apoptotic process
mitochondrion morphogenesis
cellular response to organic substance
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
intrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum unfolded protein response
regulation of mitochondrial membrane permeability involved in apoptotic process
positive regulation of mitochondrial membrane permeability involved in apoptotic process
apoptotic process involved in patterning of blood vessels
apoptotic process involved in embryonic digit morphogenesis
regulation of mitochondrial membrane permeability involved in programmed necrotic cell death
positive regulation of apoptotic DNA fragmentation
retinal cell apoptotic process
B cell receptor apoptotic signaling pathway
negative regulation of apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of intrinsic apoptotic signaling pathway
protein binding
lipid binding
channel activity
heat shock protein binding
protein complex binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
chaperone binding
BH domain binding
BH3 domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142582 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11800 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P015183 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P015183 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11800 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P015183 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11800 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11800 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700601025 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56605 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700601025 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10738462 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10721836 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10721839 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10721838 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10721840 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10721837 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10721835 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738461 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10721834 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10738460 | - | - |
| Rat Genome 230 2.0 Array | 1369122_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369122_at | - | - |
| Affymetrix HT Rat Focus Array | 1369122_at | - | - |
| Affymetrix HT Rat Focus Array | 1369122_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100044029.rn.1 | TC0100005392.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100044024.rn.1 | TC0100005392.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100044031.rn.1 | TC0100005392.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100044036.rn.1 | TC0100005392.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100044026.rn.1 | TC0100005392.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100044037.rn.1 | TC0100005392.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100044033.rn.1 | TC0100005392.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100044027.rn.1 | TC0100005392.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369122_PM_at | - | - |
| Rat Expression Set 230 | 1369122_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738460 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10721834 | - | - |
| Rat Genome U34 Set | U59184_at | - | - |
| Rat Genome U34 Set | U49729_at | - | - |
| Rat Genome U34 Set | S76511_s_at | - | - |
| Rat Genome U34 Set | U49729_at | - | - |
| Rat Genome U34 Set | U59184_at | - | - |
| Rat Genome U34 Set | S76511_s_at | - | - |
| Rat Genome U34 Set | U49729_at | - | - |
| Rat Genome U34 Set | S76511_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22354960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353843 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62519 | - | - |

Bax
Genomic Map