Assay Details
| Gene Symbol: Prrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108482.1 | NP_001101952.1 | 3 - 4 | 461 | 107 | |
| XM_006229098.2 | XP_006229160.1 | 3 - 4 | 1119 | 107 | ||
| XM_008759396.1 | XP_008757618.1 | 4 - 5 | 549 | 107 | ||
| XM_008759397.1 | XP_008757619.1 | 3 - 4 | 410 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prrg2
Genomic Map