Assay Details
| Gene Symbol: Srrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249579.2 | XP_006249641.1 | 3 - 4 | 479 | 146 | |
| XM_213799.8 | XP_213799.6 | 3 - 4 | 479 | 146 | ||
More Information
Set Membership: |
|

Srrd
Genomic Map