Assay Details
| Gene Symbol: Dtx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751193.3 | XP_003751241.1 | 3 - 4 | 1796 | 64 | |
| XM_006249412.2 | XP_006249474.1 | 3 - 4 | 1796 | 64 | ||
More Information
Set Membership: |
|

Dtx1
Genomic Map