Assay Details
| Gene Symbol: LOC100360940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729540.3 | XP_002729586.2 | 2 | 468 | 87 | |
| XM_008764180.1 | XP_008762402.1 | 1 | 255 | 87 | ||
| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008292.1 | NP_001008293.1 | 4 - 5 | 438 | 87 | |
| GenBank mRNA | BC086366.1 | - | 4 - 5 | 438 | 7305418 | 87 |
| FQ219826.1 | - | 4 - 5 | 431 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100360940
Genomic Map