Assay Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107153.1 | NP_001100623.1 | 4 - 5 | 599 | 67 | |
| XM_008769337.1 | XP_008767559.1 | 4 - 5 | 607 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp30
Genomic Map