Assay Details
| Gene Symbol: LOC100910540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751154.3 | XP_003751202.1 | 2 - 3 | 2474 | 78 | |
| Gene Symbol: Mepce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100678.1 | NP_001094148.1 | 2 - 3 | 2394 | 78 | |
| GenBank mRNA | BC088435.1 | - | 2 - 3 | 812 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910540
Genomic Map