Assay Details
| Gene Symbol: Orai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170403.1 | NP_001163874.1 | 1 - 2 | 180 | 94 | |
| GenBank mRNA | BC158835.1 | - | 1 - 2 | 84 | 7384003 | 94 |
| EU644750.1 | - | 1 - 2 | 180 | 94 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC158835.1 transcripts.
Set Membership: |
|

Orai2
Genomic Map