Assay Details
| Gene Symbol: Psmd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130430.1 | NP_569114.1 | 3 - 4 | 946 | 75 | |
| GenBank mRNA | AF067728.1 | - | 3 - 4 | 946 | 75 | |
| FQ214178.1 | - | 490 | 75 | |||
| FQ225583.1 | - | 479 | 75 | |||
| FQ227319.1 | - | 479 | 75 | |||
| Gene Symbol: LOC103694906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773903.1 | XP_008772125.1 | 2 - 3 | 683 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmd9
Genomic Map