Assay Details
| Gene Symbol: Wsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007616.1 | NP_001007617.1 | 3 - 4 | 493 | 96 | |
| XM_008769220.1 | XP_008767442.1 | 2 - 3 | 430 | 96 | ||
| GenBank mRNA | BC081901.1 | - | 3 - 4 | 493 | 7110808 | 96 |
More Information
Updated Mapping Information
Set Membership: |
|

Wsb2
Genomic Map