Assay Details
Gene Symbol: Trim50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181080.1 | NP_851594.1 | 4 - 5 | 743 | 127 | |
XM_006249129.2 | XP_006249191.1 | 5 - 6 | 1065 | 127 | ||
GenBank mRNA | AY081950.1 | - | 4 - 5 | 743 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
