Assay Details
| Gene Symbol: Muc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769185.1 | XP_008767407.1 | 5 - 6 | 616 | 132 | |
| GenBank mRNA | U76551.1 | - | 132 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Muc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769185.1 | XP_008767407.1 | 5 - 6 | 616 | 132 | |
| GenBank mRNA | U76551.1 | - | 132 | |||
Set Membership: |
|