Assay Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033684.1 | NP_001028856.1 | 10 - 11 | 1236 | 116 | |
| GenBank mRNA | BC091282.1 | - | 4 - 5 | 358 | 116 | |
| BC104703.1 | - | 10 - 11 | 1236 | 7300674 | 116 | |
| FQ216833.1 | - | 10 - 11 | 1254 | 116 | ||
| FQ218432.1 | - | 10 - 11 | 1283 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cct6a
Genomic Map