Assay Details
| Gene Symbol: Wbscr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025112.1 | NP_001020283.1 | 9 - 10 | 1504 | 98 | |
| XM_008769132.1 | XP_008767354.1 | 9 - 10 | 1372 | 98 | ||
| GenBank mRNA | AB040671.1 | - | 9 - 10 | 1504 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wbscr17
Genomic Map