Assay Details
| Gene Symbol: Wbscr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025112.1 | NP_001020283.1 | 2 - 3 | 422 | 112 | |
| XM_008769132.1 | XP_008767354.1 | 2 - 3 | 290 | 112 | ||
| GenBank mRNA | AB040671.1 | - | 2 - 3 | 422 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wbscr17
Genomic Map