Assay Details
| Gene Symbol: LOC690085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769276.1 | XP_008767498.1 | 3 - 4 | 214 | 80 | |
| GenBank mRNA | AY724520.1 | - | 2 - 3 | 292 | 80 | |
More Information
Set Membership: |
|

LOC690085
Genomic Map