Assay Details
Gene Symbol: LOC690085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769276.1 | XP_008767498.1 | 4 - 5 | 319 | 113 | |
GenBank mRNA | AY724520.1 | - | 3 - 4 | 397 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
