Assay Details
Gene Symbol: LOC690085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769276.1 | XP_008767498.1 | 5 - 6 | 482 | 61 | |
GenBank mRNA | AY724520.1 | - | 4 - 5 | 560 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
