Assay Details
| Gene Symbol: Nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251935.2 | XP_006251997.1 | 2 - 3 | 244 | 146 | |
| XM_006251937.2 | XP_006251999.1 | 2 - 3 | 224 | 146 | ||
| XM_008770672.1 | XP_008768894.1 | 1 - 2 | 147 | 146 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006251935.2, XM_006251937.2, XM_008770672.1 transcripts.
Set Membership: |
|

Nrl
Genomic Map