Assay Details
| Gene Symbol: Defb39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037522.1 | NP_001032611.1 | 1 - 2 | 128 | 124 | |
| GenBank mRNA | AY621366.1 | - | 1 - 2 | 128 | 124 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY621366.1, NM_001037522.1 transcripts.
Set Membership: |
|

Defb39
Genomic Map