Assay Details
| Gene Symbol: Tubgcp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107323.1 | NP_001100793.1 | 18 - 19 | 2231 | 66 | |
| XM_006253440.2 | XP_006253502.1 | 19 - 20 | 2395 | 66 | ||
| XM_006253441.2 | XP_006253503.1 | 18 - 19 | 2287 | 66 | ||
| GenBank mRNA | BC085356.1 | - | 4 - 5 | 378 | 66 | |
| FQ222809.1 | - | 21 - 22 | 2589 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp3
Genomic Map