Assay Details
| Gene Symbol: Adam3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020302.1 | NP_064698.1 | 15 - 16 | 1732 | 159 | |
| GenBank mRNA | BC078839.1 | - | 15 - 16 | 1638 | 7114315 | 159 |
| Y07903.1 | - | 15 - 16 | 1732 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam3a
Genomic Map