Assay Details
| Gene Symbol: Unc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107319.2 | NP_001100789.2 | 5 - 6 | 746 | 64 | |
| XM_008771325.1 | XP_008769547.1 | 2 - 3 | 239 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Unc5d
Genomic Map