Assay Details
| Gene Symbol: Pomk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024883.1 | NP_001020054.1 | 3 - 4 | 280 | 68 | |
| GenBank mRNA | BC097466.1 | - | 3 - 4 | 280 | 7457667 | 68 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC097466.1,NM_001024883.1 transcripts.
Set Membership: |
|

Pomk
Genomic Map