Assay Details
| Gene Symbol: Dctn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106085.1 | NP_001099555.1 | 4 - 5 | 363 | 90 | |
| GenBank mRNA | FQ212035.1 | - | 4 - 5 | 349 | 90 | |
| FQ226689.1 | - | 4 - 5 | 386 | 90 | ||
| FQ233791.1 | - | 4 - 5 | 380 | 90 | ||
| FQ234632.1 | - | 4 - 5 | 386 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dctn6
Genomic Map