Assay Details
| Gene Symbol: Tm2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017444.1 | NP_001017444.1 | 1 - 2 | 301 | 124 | |
| GenBank mRNA | BC093382.1 | - | 1 - 2 | 301 | 7389050 | 124 |
More Information
Updated Mapping Information
Set Membership: |
|

Tm2d2
Genomic Map