Assay Details
| Gene Symbol: Coprs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169121.1 | NP_001162592.1 | 3 - 4 | 373 | 59 | |
| GenBank mRNA | CB614079.1 | - | 3 - 4 | 373 | 59 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Coprs
Genomic Map